Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD133 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB2016518SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB2016518SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB2016518SS Supplier Novus Biologicals Supplier No. NB12016518SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 26 publications

CD133 Polyclonal specifically detects CD133 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CD133
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Flow Cytometry 1:10-1:1000, ELISA 1:1000, Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100-1:250, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP)
Formulation PBS with 0.02% Sodium Azide
Gene Alias AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1
Gene Symbols PROM1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a C-terminal region of CD133 (within amino acids 750-865).
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Core ESC Like Genes, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8842
Test Specificity CD133 - Hematopoietic Stem Cell Marker
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.