Learn More
Invitrogen™ CD133 (Prominin-1) Monoclonal Antibody (TMP4), Alexa Fluor™ 488, eBioscience™

Description
Description: The TMP4 monoclonal antibody reacts with human CD133 (Prominin-1), a 120 kDa member of the pentaspan family of proteins, which also includes Prominin-2. Their expression is found within plasma membrane protrusions such as epithelial microvilli. CD133 can exist in a number of alternatively spliced isoforms, and the protein has several N-linked glycosylation sites: the occurrence of both may be tissue-dependent. Human CD133 was first identified as an epitope expressed on CD34+ hematopoietic progenitors. Although the ligand and function of CD133 remain unknown, it has since proven to be very useful as a marker for both stem cells and cancer stem cells. In addition to its expression on hematopoietic precursors, CD133 has been used to identify tumorigenic colon cancer stem cells, brain cancer stem cells, prostate cancer stem cells, in addition to others. The binding of the TMP4 antibody does not block the binding of another anti-human CD133 antibody, EMK08 indicating that they recognize distinct epitopes. Applications Reported: This TMP4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TMP4 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can r...
Specifications
Specifications
| Antigen | CD133 (Prominin-1) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | TMP4 |
| Concentration | 5 μL/Test |
| Conjugate | Alexa Fluor 488 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | PROM1 |
| Gene Accession No. | O43490 |
| Gene Alias | 4932416E19Rik; AC133; Antigen AC133; antigen AC133 homolog; antigen CD133; CD107b; CD133; CORD12; fudenine; hematopoietic stem cell antigen; hProminin; Lamp II; Lamp2; LAMP-2; Lamp-2a; Lamp-2b; Lamp-2c; LGP-96; LGP-B; Mac3; MCDR2; MSTP061; Prom; PROM1; Prom-1; prominin 1; prominin 1.s1; Prominin1; prominin-1; prominin-1.s2; prominin-like 1; prominin-like protein 1; PROML1; RP41; STGD4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.