Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Antibody (433423), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB347767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347767 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347767 Supplier Novus Biologicals Supplier No. FAB4597MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD14 Monoclonal specifically detects CD14 in Equine, Pig samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen CD14
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 433423
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
Host Species Mouse
Immunogen E. coli-derived recombinant porcine CD14
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 929
Target Species Equine, Pig
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.