Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Antibody (CL1638), Novus Biologicals™
SDP

Catalog No. NB436554 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB436554 25 μL
NBP234486 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB436554 Supplier Novus Biologicals Supplier No. NBP23448625UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CD14
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL1638
Conjugate Unconjugated
Dilution Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin
Gene Accession No. D6RFL4
Gene Alias CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
Gene Symbols CD14
Host Species Mouse
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: PCELDDEDFRCVCNFSEPQPDWSEAFQCVSAVEVEIHAGGLNLEPFLKRVDADADPRQYADTVKALRVRRLTVGAAQVPAQLLVGALRVLAYSRLKELTLEDLKITGTMPPLPLEATGLALSSL
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Asthma, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 929
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.