Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD14 Antibody (CL1638), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $646.00
Specifications
Antigen | CD14 |
---|---|
Clone | CL1638 |
Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CD14 | |
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
CD14 | |
IgG1 | |
Protein A purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL1638 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Apoptosis, Asthma, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers | |
D6RFL4 | |
929 | |
This antibody was developed against a recombinant protein corresponding to amino acids: PCELDDEDFRCVCNFSEPQPDWSEAFQCVSAVEVEIHAGGLNLEPFLKRVDADADPRQYADTVKALRVRRLTVGAAQVPAQLLVGALRVLAYSRLKELTLEDLKITGTMPPLPLEATGLALSSL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title