Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Antibody (RPA-M1), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP22721A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP22721A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP22721A70 Supplier Novus Biologicals Supplier No. NBP227215AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CD14
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone RPA-M1
Conjugate Alexa Fluor 750
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
Gene Symbols CD14
Host Species Mouse
Immunogen Human PHA-stimulated PBMC were used as immunogen.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 929
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.