Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Antibody (TUK4), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB10062994S Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062994S 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062994S Supplier Novus Biologicals Supplier No. NB10062994R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for ELISA, Flow Cytometry.

Specifications

Antigen CD14
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone TUK4
Conjugate DyLight 550
Dilution Flow Cytometry, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
Gene Symbols CD14
Host Species Mouse
Immunogen Made to Human CD14
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 929
Test Specificity This product recognises the human CD14 cell surface antigen, expressed strongly by most peripheral blood monocytes and weakly on granulocytes. Clone Tuk4 has been shown to block SDF-induced chemotaxis of U937 cells in a dose-dependent manner(6).
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.