Learn More
Description
Specifications
Specifications
| Antigen | CD140a |
| Applications | Single Cell 3' Sequencing |
| Classification | Monoclonal |
| Clone | Clone APA5 |
| Conjugate | Oligo |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | Pdgfra; Pdgfr2; PDGF-R alpha; Platelet derived growth factor receptor alpha |
| Host Species | Rat |
| Immunogen | Mouse PDGF Receptor α chain |
| Quantity | 25 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.