Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD15/Lewis X Mouse anti-Human, Clone: ARE, Novus Biologicals™
SDP

Catalog No. NB151714 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151714 25 μg
NB151713 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151714 Supplier Novus Biologicals Supplier No. NBP31188025UG

Mouse Monoclonal Antibody

CD15/Lewis X Monoclonal antibody specifically detects CD15/Lewis X in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CD15/Lewis X
Applications Flow Cytometry
Classification Monoclonal
Clone ARE
Conjugate Unconjugated
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD 15, CD-15, Lewis X, Lewis-X, LeX, SSEA-1, stage-specific embryonic antigen 1
Host Species Mouse
Immunogen Human CD15/Lewis X (Uniprot: P22083)
Purification Method Protein A or G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2526
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.