Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ CD15, Mouse Monoclonal Antibody, 200μg/mL; Unlabeled; Purified with BSA and azide

Obtain accurate and reproducible results in immunohistochemistry applications with Epredia™ CD15, Mouse Monoclonal Antibody.

Specifications

Antigen CD15
Clone SPM490
Applications Immunohistochemistry
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MS10338P0
View Documents
Epredia™
MS10338P0
100 μL N/A N/A N/A
MS10338P1
View Documents
Epredia™
MS10338P1
500 μL N/A N/A N/A
Description

Description

CD15 or X-hapten plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. CD15 is also expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin's disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.

Host Species: Mouse

Clone: SPM490

Isotype: IgM

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: K562 (an erythroid-myeloid leukemia cell line)

Molecular Weight: 220kDa

Positive Control: Reed-Sternberg's cells in Hodgkin's lymphoma

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD15
Immunohistochemistry
Unconjugated
Human
IgM
Reed-Sternberg's cells in Hodgkin's lymphoma
SPM490
Monoclonal
Mouse
K562 (an erythroid-myeloid leukemia cell line)
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.