Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD151 Antibody (11G5a), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB121043 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB121043 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB121043 Supplier Novus Biologicals Supplier No. NBP308797AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD151 Monoclonal antibody specifically detects CD151 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CD151
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 11G5a
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, tetraspanin-24, tspan-24, TSPAN24hemidesmosomal tetraspanin CD151
Host Species Mouse
Immunogen The details of the immunogen for this antibody are not available.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 977
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.