Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD151 Antibody (B-E31), HRP, Novus Biologicals™
SDP

Catalog No. NB340631 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340631 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340631 Supplier Novus Biologicals Supplier No. NBP318100H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD151 Monoclonal antibody specifically detects CD151 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CD151
Applications Flow Cytometry
Classification Monoclonal
Clone B-E31
Conjugate HRP
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, tetraspanin-24, tspan-24, TSPAN24hemidesmosomal tetraspanin CD151
Host Species Mouse
Immunogen Eahy 926 cell line. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cell
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 977
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.