Learn More
MilliporeSigma™ CD152 (CTLA-4), Mouse, PE, Clone: BNI3,
Mouse Monoclonal Antibody
Specifications
Antigen | CD152 (CTLA-4) |
---|---|
Clone | BNI3 |
Concentration | Please refer to lot specific datasheet. |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The BNI3 antibody is specific for human CD152, commonly known as CTLA-4, a 33-37kDa protein expressed as a homodimer on the surface of activated T and B cells, and on thymocytes. CTLA-4 is structurally similar, yet functionally disparate, to the T cell co-stimulatory molecule CD28. Both CTLA-4 and CD28 interact with the co-stimulatory molecules CD80 (B7-1) and CD86 (B7-2) on antigen-presenting cells, with CTLA-4 displaying a higher avidity than CD28. While CD28 typically delivers a potent co-stimulatory signal in support of T cell activation, CTLA-4 appears to act as a negative regulator of T cell activation and may contribute to the suppressor function of Treg cells.CTLA-4 proteins may be initially sequestered within Golgi vesicles, from which they can be transferred to and from the cell surface, a mechanism by which Treg cells can selectively impart suppressive functions. The BNI3 antibody may be used for flow cytometric analysis of intracellular or surface CTLA-4 expression, and is also widely used for neutralization of CTLA-4 when expressed at the cell surface. The BNI3 antibody is reported to be cross-reactive with Baboon, Cynomolgus and Rhesus CTLA-4. Please choose the appropriate format for each application.
Specifications
CD152 (CTLA-4) | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P16410 | |
IgG2a | |
Purified by size-exclusion chromatography |
BNI3 | |
Flow Cytometry | |
PE | |
Mouse | |
Inflammation & Immunology | |
Purified mouse monoclonal IgG2a in buffer containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2. | |
Immunogen information for this antibody is proprietary information. | |
Primary | |
CTLA4, CD152 |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.