Learn More
CD152 Hamster anti-Mouse, PE, Clone: UC10-4F10-11, BD
Hamster Monoclonal Antibody
Supplier: BD Biosciences 561718
Description
The UC10-4F10-11 antibody reacts with CD152 (CTLA-4), which is expressed on activated T lymphocytes 2-3 days after stimulation through T cell receptor. CTLA-4 has significant similarity to CD28 in amino acid sequence, structure, and genomic organization. Furthermore, CD152 and CD28 share common B7 family counter-receptors. Unlike CD28, CD152 expression appears to be restricted to activated T cells and CD25+CD4+ regulatory T (Treg) cells. Whereas CD28 delivers a costimulatory signal required for T-cell activation, CTLA-4 is a negative regulator of cell-mediated immune responses. CD152 may play roles in induction and/or maintenance of immunological tolerance, regulation of protective immunity, and autoimmune responses, and regulation of some aspects of thymocyte maturation. This hamster mAb to a mouse leukocyte antigen does not cross-react with rat leukocytes.

Specifications
CD152 | |
Monoclonal | |
0.2mg/mL | |
CTLA-4 | |
Hamster | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
UC10-4F10-11 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse CTLA-4 IgG2a Fusion | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.