Learn More
CD152 Hamster anti-Mouse, PE, Clone: UC10-4F10-11, BD
Hamster Monoclonal Antibody
$78.00 - $252.00
Specifications
Antigen | CD152 |
---|---|
Clone | UC10-4F10-11 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561718
|
BD Biosciences
561718 |
25 μg |
Each for $78.00
|
|
|||||
BDB553720
|
BD Biosciences
553720 |
0.1 mg |
Each for $252.00
|
|
|||||
Description
The UC10-4F10-11 antibody reacts with CD152 (CTLA-4), which is expressed on activated T lymphocytes 2-3 days after stimulation through T cell receptor. CTLA-4 has significant similarity to CD28 in amino acid sequence, structure, and genomic organization. Furthermore, CD152 and CD28 share common B7 family counter-receptors. Unlike CD28, CD152 expression appears to be restricted to activated T cells and CD25+CD4+ regulatory T (Treg) cells. Whereas CD28 delivers a costimulatory signal required for T-cell activation, CTLA-4 is a negative regulator of cell-mediated immune responses. CD152 may play roles in induction and/or maintenance of immunological tolerance, regulation of protective immunity, and autoimmune responses, and regulation of some aspects of thymocyte maturation. This hamster mAb to a mouse leukocyte antigen does not cross-react with rat leukocytes.
Specifications
CD152 | |
0.2mg/mL | |
Monoclonal | |
Hamster | |
Mouse | |
Mouse CTLA-4 IgG2a Fusion | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
UC10-4F10-11 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
CTLA-4 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.