Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD155 Monoclonal Antibody (2H7CD155), PerCP-eFluor™ 710, eBioscience™
GREENER_CHOICE

Catalog No. p-7168691
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-9165 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-9165 Supplier Invitrogen™ Supplier No. 46155042
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The monoclonal antibody 2H7CD155 recognizes CD155 also known as the poliovirus receptor. The CD155 protein is a member of the Ig superfamily in which 4 isoforms exist; only the alpha and delta forms are expressed on the membrane of monocytes, prostate cancer cells, as well as neurectodermal and monocytic tumor cells lines. In addition, CD155 is found during mid-gastrulation during embryonic development. CD155 is thought to play a role in adhesion by interaction with the ECM component vitronectin as well as a role in NK killing of tumor cells. CD226 and CD96 are ligands for CD155. Blocking of CD155 is under therapeutic investigation for prevention of variants of poliovirus. Applications Reported: This 2H7CD155 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2H7CD155 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells or human tumor cell line U937. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrum...

The protein encoded by this gene is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The external domain mediates cell attachment to the extracellular matrix molecule vitronectin, while its intracellular domain interacts with the dynein light chain Tctex-1/DYNLT1. The gene is specific to the primate lineage, and serves as a cellular receptor for poliovirus in the first step of poliovirus replication. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD155
Applications Flow Cytometry
Classification Monoclonal
Clone 2H7CD155
Concentration 5 μL/Test
Conjugate PerCP-eFluor 710
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene PVR
Gene Accession No. P15151
Gene Alias 3830421F03Rik; AI325026; AI987993; CD112; CD155; D7Ertd458e; FLJ25946; herpes virus entry mediator B; Herpesvirus entry mediator B; hveB; HVED; mE4; mHveB; Mph; murine herpes virus entry protein B; murine herpesvirus entry protein B; NECL5; necl-5; Nectin cell adhesion molecule 2; Nectin2; Nectin-2; nectin-like 5; nectin-like protein 5; Poliovirus receptor; poliovirus receptor homolog; poliovirus receptor-related 2; poliovirus receptor-related protein 2; poliovirus sensitivity; PVR; Pvrl2; PVS; sCD112; soluble CD112; Taa1; TAGE4; tumor-associated antigen 1; tumor-associated glycoprotein pE4
Gene Symbols PVR
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5817
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.