Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD155/PVR Antibody (114), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB088341 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB088341 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB088341 Supplier Novus Biologicals Supplier No. NBP289272C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD155/PVR Monoclonal specifically detects CD155/PVR in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CD155/PVR
Applications Western Blot, ELISA
Classification Monoclonal
Clone 114
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CD155/PVR extracellular domain (Met 1-Asn 343; NP_006496.2).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 5817
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.