Learn More
Description
Specifications
Specifications
| Antigen | CD155/PVR |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 114 |
| Conjugate | FITC |
| Dilution | ELISA |
| Formulation | PBS |
| Gene Alias | CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human CD155/PVR extracellular domain (Met 1-Asn 343; NP_006496.2). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
