Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD155/PVR Antibody (690912) [FITC], Novus Biologicals™
SDP

Catalog No. NB375511 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375511 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375511 Supplier Novus Biologicals Supplier No. FAB6909F
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD155/PVR Monoclonal antibody specifically detects CD155/PVR in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF

Specifications

Antigen CD155/PVR
Applications Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 690912
Conjugate FITC
Dilution Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias CD155, CD155 antigen, HVED, NECL5, NECL-5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD155/PVR, Asp29-Leu348, Accession # NP_081790
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 5817
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.