Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD157 Monoclonal Antibody (eBioSY11B5 (SY11B5)), NovaFluor™ Yellow 590, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ H041T03Y02A

Description
Bone marrow stromal cell antigen 1 (BST1) is a stromal cell line-derived glycosylphosphatidylinositol-anchored molecule that facilitates pre-B cell growth. BST1, which is also known as ADP-rybosyl cyclase 2 and DAMP-1, is an enzyme which is expressed by BST1 gene in human. BST1 expression is enhanced in bone marrow stromal cell lines derived from patients with rheumatoid arthritis. The polyclonal B cell abnormalities in rheumatoid arthritis may be, at least in part, attributed to BST1 overexpression in the stromal cell population.
Specifications
CD157 | |
Monoclonal | |
0.4 μg/Test | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q10588 | |
Bst1 | |
Affinity chromatography | |
RUO | |
683 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid | |
IgG1 κ |
Flow Cytometry | |
eBioSY11B5 (SY11B5) | |
NovaFluor Yellow 590 | |
Bst1 | |
114/A10; A530073F09; ADP-ribosyl cyclase 2; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 2; antigen BP3; Bone marrow stromal antigen 1; bone marrow stromal cell antigen 1; Bp3; Bp-3; BP-3 alloantigen; Bst1; BST-1; Bsta1; cADPr hydrolase 2; CD157; CD157 antigen; Cyclic ADP-ribose hydrolase 2; leukocyte antigen 65; Ly65; Ly-65; NAD(+) nucleosidase; NAD+ nucleosidase | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction