Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD157 Monoclonal Antibody (eBioSY11B5 (SY11B5)), NovaFluor™ Yellow 590, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD157 |
---|---|
Clone | eBioSY11B5 (SY11B5) |
Concentration | 0.4 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H041T03Y02A
![]() |
Invitrogen™
H041T03Y02A |
100 Tests |
Each for $430.00
|
|
|||||
H041T02Y02A
![]() |
Invitrogen™
H041T02Y02A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD157 Monoclonal antibody specifically detects CD157 in Human samples. It is validated for Flow CytometrySpecifications
CD157 | |
0.4 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
Q10588 | |
683 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
eBioSY11B5 (SY11B5) | |
Flow Cytometry | |
NovaFluor Yellow 590 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
114/A10; A530073F09; ADP-ribosyl cyclase 2; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 2; antigen BP3; Bone marrow stromal antigen 1; bone marrow stromal cell antigen 1; Bp3; Bp-3; BP-3 alloantigen; Bst1; BST-1; Bsta1; cADPr hydrolase 2; CD157; CD157 antigen; Cyclic ADP-ribose hydrolase 2; leukocyte antigen 65; Ly65; Ly-65; NAD(+) nucleosidase; NAD+ nucleosidase | |
IgG1 κ | |
Affinity chromatography | |
Bst1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title