Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16/CD32 Antibody (2.4G2), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP22644A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP22644A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP22644A30 Supplier Novus Biologicals Supplier No. NBP252644AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD16/CD32 Monoclonal specifically detects CD16/CD32 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence.

Specifications

Antigen CD16/CD32
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 2.4G2
Conjugate Alexa Fluor 350
Dilution Flow Cytometry, Immunohistochemistry, Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD16, CD16A, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), FCG3, FCGR3, FCGR3A, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3
Gene Symbols FCGR3A
Host Species Rabbit
Immunogen Mouse BALB/c Macrophage J774 Cell Line.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2214
Test Specificity This antibody binds to an epitope on the extracellular domains of the mouse Fc(gamma)III and Fc(gamma)II, thereby inhibiting their ability to bind antibodies via their Fc region. receptors. Also binding the Fc(gamma)I receptor (CD64) via its Fc domain.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.