Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16/CD32 Antibody (93), Biotin, Novus Biologicals™
SDP

Catalog No. NBP127946B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP127946B 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP127946B Supplier Novus Biologicals Supplier No. NBP127946B
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD16/CD32 Monoclonal specifically detects CD16/CD32 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen CD16/CD32
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 93
Conjugate Biotin
Dilution Immunohistochemistry-Frozen
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD16, CD16A, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), FCG3, FCGR3, FCGR3A, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3
Gene Symbols FCGR3A
Host Species Rat
Immunogen Murine CD16/32 (CD16/Fc gamma III and CD32/FcgammaII receptors)
Purification Method Protein A or G purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2214
Test Specificity Murine CD16/32 (CD16/Fc gamma III and CD32/Fc gamma II receptors)
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.