Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD160 Antibody (001), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB069553 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB069553 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB069553 Supplier Novus Biologicals Supplier No. NBP290072R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD160 Monoclonal specifically detects CD160 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen CD160
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate DyLight 550
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias BY55FLJ46513, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CD160 (NP_008984.1; Met1-Leu158).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11126
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.