Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD160 Antibody (BY55), Novus Biologicals™
SDP

Catalog No. NB018409 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018409 0.025 mg
NBP121435 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018409 Supplier Novus Biologicals Supplier No. NBP1214350.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD160 Monoclonal specifically detects CD160 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.

Specifications

Antigen CD160
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone BY55
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:50, Immunoprecipitation 1:10-1:500
Formulation PBS (pH 7.4) with 0.09% Sodium Azide
Gene Accession No. O95971
Gene Alias BY55FLJ46513, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55
Gene Symbols CD160
Host Species Mouse
Immunogen Human NK cell line YT2C2
Purification Method IgM purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11126
Test Specificity NBP1-21435 recognizes human CD160, a 27kD cell surface glycoprotein that was initially identified by clone BY55. The expression of CD160 is restricted to circulating NK cells, lymphocytes expressing T-cell receptor (TCR) gamma/delta and a small population of TCR alpha/beta positive T cells. In tissues, CD160 is expressed by intestinal intraepithelial lymphocytes.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.