Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD160 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB200078 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200078 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200078 Supplier Novus Biologicals Supplier No. NBP241234F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD160 Polyclonal antibody specifically detects CD160 in Human, Mouse samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen CD160
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias BY55FLJ46513, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid peptide near the center of human CD160. The immunogen is located within amino acids 50 - 100 of CD160 . Amino Acid Squence: SPETSLKQLRLKRDPG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11126
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.