Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD161 Antibody (10/78), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB10065297A Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065297A 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065297A Supplier Novus Biologicals Supplier No. NB10065297R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD161 Monoclonal specifically detects CD161 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.

Specifications

Antigen CD161
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 28764
Conjugate DyLight 550
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR
Gene Symbols KLRB1
Host Species Mouse
Immunogen Purified splenic NK cells from the LEW rat strain.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3820
Test Specificity Recognizes the rat CD161 protein (also known as NKR-PI) a 60kD glycoprotein expressed on rat NK cells and T cell subpopulations. This competes with clone 3.2.3 for binding to antigen.
Target Species Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.