Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD161 Antibody (14F2F11), Novus Biologicals™
SDP

Catalog No. NBP214844 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP214844 0.1 mg
NBP214844SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP214844 Supplier Novus Biologicals Supplier No. NBP214844
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Western Blot.

Specifications

Antigen CD161
Applications Western Blot
Classification Monoclonal
Clone 14F2F11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000
Gene Accession No. Q12918
Gene Alias CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR
Gene Symbols KLRB1
Host Species Mouse
Immunogen Amino acids 68-225 of human CD161 were used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3820
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.