Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD161 Antibody (14F2F11), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $426.00
Specifications
Antigen | CD161 |
---|---|
Clone | 14F2F11 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1:100 - 1:2000 |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP214844SS
![]() |
Novus Biologicals
NBP214844SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP214844
![]() |
Novus Biologicals
NBP214844 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Western Blot.Specifications
CD161 | |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
Signal Transduction | |
Q12918 | |
3820 | |
Amino acids 68-225 of human CD161 were used as the immunogen for this antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
14F2F11 | |
Western Blot 1:100 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
KLRB1 | |
IgG2b κ | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title