Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD161 Antibody (B199.2), PerCP, Novus Biologicals™
SDP

Catalog No. NB10065298P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065298P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065298P Supplier Novus Biologicals Supplier No. NB10065298PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen CD161
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot (Negative)
Classification Monoclonal
Clone B199.2
Conjugate PerCP
Dilution Flow Cytometry, Western Blot (Negative)
Gene Accession No. Q12918
Gene Alias CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR
Gene Symbols KLRB1
Host Species Mouse
Immunogen NB100-65298 recognises the human CD161 cell surface antigen, expressed by almost all NK cells and with a small subset of CD3+ve T cells.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3820
Test Specificity CD161 (B199.2)
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.