Learn More
CD161 Mouse anti-Human, PE, Clone: DX12, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 340530
Description
CD161, clone DX12, is derived from splenocytes from C3H/HeJ mice immunized with murine fibroblasts stably transfected with hNKR-P1A cDNA in the pBJ expression vector and fused with Sp2/0 myeloma cells.
CD161 recognizes the human NKR-P1A antigen, a disulfide-linked homodimer of a 40-kdalton (kDa) glycoprotein reduced to 26 kDa by treatment with N-glycanase.

Specifications
CD161 | |
Monoclonal | |
PE | |
Mouse | |
50 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
DX12 | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Human NKR-P1A Transfected Mouse Fibroblasts | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.