Learn More
CD161 Mouse anti-Human, PE, Clone: DX12, BD
Mouse Monoclonal Antibody
$306.00 - $331.50
Specifications
Antigen | CD161 |
---|---|
Clone | DX12 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB340530
|
BD Biosciences
340530 |
50 Tests |
Each for $331.50
|
|
|||||
BDB556081
|
BD Biosciences
556081 |
100 Tests |
Each for $306.00
|
|
|||||
Description
The DX12 monoclonal antibody specifically binds to human CD161 which is also known as Natural killer cell surface protein P1A (NKR-P1A). CD161 is encoded by the KLRB1 (Killer cell lectin-like receptor subfamily B member 1) gene. CD161 is a member of the C-type lectin superfamily and is also referred to as C-type lectin domain family 5 member B (CLEC5B). CD161 is a 80 kDa disulfide-linked homodimer, type II membrane glycoprotein. CD161 is expressed mostly on NK cell populations and on subsets of CD4+ and CD8+ αβ T cells, NKT cells and γδ T cells. Reports indicate that CD161 is expressed preferentially on CD45RO+ T cells, however, it can be found on a subset of thymocytes and fetal liver T cells. Its function has not been fully elucidated, but reports indicate that NKR-P1A may serve as a specific receptor for some NK cell targets. The DX12 antibody can reportedly inhibit spontaneous cytotoxicity mediated by certain NK cell clones.
Specifications
CD161 | |
Flow Cytometry | |
PE | |
RUO | |
Human | |
IgG1 κ |
DX12 | |
Monoclonal | |
Mouse | |
Immunology | |
Human NKR-P1A Transfected Mouse Fibroblasts | |
Primary |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.