Learn More
Description
Specifications
Specifications
| Antigen | CD161/NK1.1 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | PK136 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry < 1.0 ug/10^6 cells, Immunohistochemistry 1:20-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:1000, Immunoprecipitation 1:20-1:1000, Immunohistochemistry-Paraffin 1:20-1:1000. Use reported in scientific literature (PMID 21118981), Immunohistochemistry-Frozen 1:20-1:1000 |
| Gene Alias | CD161, CD161 antigen, CD161c, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR |
| Gene Symbols | Klrb1c |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
