Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD161/NK1.1 Antibody (PK136) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 6 publications

$222.20 - $549.92

Specifications

Antigen CD161/NK1.1
Clone PK136
Concentration 1.0 mg/mL
Dilution Flow Cytometry < 1.0 ug/10^6 cells, Immunohistochemistry 1:20-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:1000, Immunoprecipitation 1:20-1:1000, Immunohistochemistry-Paraffin 1:20-1:1000. Use reported in scientific literature (PMID 21118981), Immunohistochemistry-Frozen 1:20-1:1000
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10077528T
SDP
View Documents
Novus Biologicals
NB10077528SS
0.1 mg
Each for $222.20
Only null left
Add to Cart
 
NB10077528
SDP
View Documents
Novus Biologicals
NB10077528
0.5 mg
Each for $549.92
Only null left
Add to Cart
 
Description

Description

CD161/NK1.1 Monoclonal specifically detects CD161/NK1.1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

CD161/NK1.1
1.0 mg/mL
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Mouse
17059
NK-1+ cells from mouse spleen and bone marrow
Primary
Store at 4C. Do not freeze.
PK136
Flow Cytometry < 1.0 ug/10^6 cells, Immunohistochemistry 1:20-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:1000, Immunoprecipitation 1:20-1:1000, Immunohistochemistry-Paraffin 1:20-1:1000. Use reported in scientific literature (PMID 21118981), Immunohistochemistry-Frozen 1:20-1:1000
Monoclonal
Mouse
Signal Transduction
CD161, CD161 antigen, CD161c, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR
Klrb1c
IgG2a κ
Affinity Purified
Mouse Monoclonal to NK-1.1 (PK136)
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.