Learn More
CD162 (PSGL-1) Monoclonal Antibody (4RA10), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$196.00 - $453.00
Specifications
Antigen | CD162 (PSGL-1) |
---|---|
Clone | 4RA10 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
53-162-180
![]() |
Invitrogen™
53162180 |
25 μg |
Each for $196.00
|
![]() |
|||||
53-162-182
![]() |
Invitrogen™
53162182 |
100 μg |
Each for $453.00
|
![]() |
|||||
Description
Description: This 4RA10 monoclonal antibody reacts with mouse CD162, which is also known as P-selectin glycoprotein ligand-1 (PSGL-1). This clone has been successfully used in flow cytometry and has been reported to block binding of CD162 to CD62P and CD62L. Applications Reported: This 4RA10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 4RA10 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser
CD162 (PSGL-1) is a 120 kDa homodimer protein found on the surface of leukocytes and platelets. CD162 is known to bind CD62P (P-selectin), CD62L (L-selectin), and to a lower degree CD62E (E-selectin). These interactions play a critical role in leukocyte adhesion, rolling and extravasation. CD162 has been shown to be an important immune marker of T cell exhaustion in chronic viral infections and cancer.Specifications
CD162 (PSGL-1) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG | |
SELPLG | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
4RA10 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Rat | |
Mouse | |
Q62170 | |
20345 | |
Mouse PSGL-1 human IgG1 fusion protein. | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
SELPLG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.