Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD163 Antibody (6D3.2F3), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB006958 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB006958 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB006958 Supplier Novus Biologicals Supplier No. NBP236495FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CD163
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6D3.2F3
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
Gene Symbols CD163
Host Species Mouse
Immunogen A synthetic peptide made to the human CD163 protein sequence (between residues 200-400). [Swiss-Prot: Q86VB7]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Test Specificity This antibody is specific for the extracellular domain of human CD163 protein
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.