Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD163 Antibody (ED2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB002079 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002079 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002079 Supplier Novus Biologicals Supplier No. NBP239099100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

CD163 Monoclonal specifically detects CD163 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CD163
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ED2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:100, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:100, Immunohistochemistry-Frozen 1:50 - 1:100
Formulation Phosphate buffered saline
Gene Accession No. Q86VB7
Gene Alias CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
Gene Symbols CD163
Host Species Mouse
Immunogen Rat Spleen cell homogenate. Spleen cells from immunised BALB/c mice were fused with cells of the SP2/0-Ag 14 mouse myeloma cell line.
Quantity 100 μg
Research Discipline Immune System Diseases, Immunology, Innate Immunity
Gene ID (Entrez) 9332
Test Specificity This antibody recognizes the rat ED2 cell surface glycoprotein.
Target Species Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.