Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD163 Antibody (EDHu-1), HRP, Novus Biologicals™
SDP

Catalog No. NB003878 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003878 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003878 Supplier Novus Biologicals Supplier No. NB11040686H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD163 Monoclonal specifically detects CD163 in Human, Rat, Porcine, Bovine, Guinea Pig, Monkey, Primate, Rhesus Macaque, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CD163
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone EDHu-1
Conjugate HRP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS with No Preservative
Gene Alias CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
Gene Symbols CD163
Host Species Mouse
Immunogen Leucocytes harvested from pleural cavity of patients with idiopathic spontaneous pneumothorax.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Test Specificity NB110-40686 recognizes the human CD163 cell surface antigen, a 130-140kD glycoprotein expressed by tissue macrophages. CD163 is not expressed by resting peripheral blood leucocytes, but expression may be induced on monocytes by culture in dexamethasone.
Target Species Human, Pig, Bovine, Guinea Pig, Monkey, Primate, Rhesus Monkey, Sheep
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.