Learn More
CD163 Antibody (GHI/61), Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP1433410.1MG
 View more versions of this product
Catalog No. NBP14334101
Only null left
Add to Cart
Description
Description
CD163 Monoclonal specifically detects CD163 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
CD163
Monoclonal
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
CD163
The immunogen for this antibody was CD163.
0.1 mg
Primary
Human
IgG1 κ
Western Blot, Flow Cytometry
GHI/61
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
Mouse
Affinity Purified
RUO
9332
Store at 4C. Do not freeze.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only