Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD163 Antibody (GHI/61), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$501.50

Specifications

Antigen CD163
Clone GHI/61
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000
Applications Western Blot, Flow Cytometry
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP14334101
SDP
View Documents
Novus Biologicals
NBP1433410.1MG
0.1 mg
Each of 1 for $501.50
Only null left
Add to Cart
 
Description

Description

CD163 Monoclonal specifically detects CD163 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

CD163
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000
Monoclonal
Mouse
Human
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
CD163
IgG1 κ
Affinity Purified
GHI/61
Western Blot, Flow Cytometry
Unconjugated
RUO
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
9332
The immunogen for this antibody was CD163.
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.