Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD163 Antibody (K20-T), Novus Biologicals™
SDP

Catalog No. NBP13014801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP13014801 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP13014801 Supplier Novus Biologicals Supplier No. NBP1301480.1ML
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody has been used in 7 publications

CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CD163
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone K20-T
Concentration 2.mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100-200, Immunohistochemistry-Frozen 1:10-1:500
Formulation 20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. Q86VB7
Gene Alias CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
Gene Symbols CD163
Host Species Rabbit
Immunogen Peptide derived from N-terminal sequence of human CD163.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9332
Test Specificity This antibody is specific for the N terminus of CD163
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.