Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD163 Antibody (R20) [DyLight 550], Novus Biologicals™
SDP

Catalog No. NB399352 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399352 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399352 Supplier Novus Biologicals Supplier No. FAB16072L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen CD163
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone R20
Conjugate DyLight 550
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163
Host Species Mouse
Immunogen Balb/c mice immunized with Synovial cells from Rheumatoid Arthritis patients
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 9332
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.