Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD163 Mouse anti-Human, Alexa Fluor™ 488, Clone: MAC2-158 (also known as Mac 2-158; Mac-2–158), BD Pharmingen™
Mouse Monoclonal Antibody
$148.00 - $329.50
Specifications
Antigen | CD163 |
---|---|
Clone | MAC2-158 (also known as Mac 2-158; Mac-2–158) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 488 |
Description
CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Flow cytometry (Routinely Tested)Specifications
CD163 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
CD163; HbSR; M130;MM130;SCARI1;SR-I1 | |
IgG1 κ | |
Affinity Chromatography |
MAC2-158 (also known as Mac 2-158; Mac-2–158) | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Monocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title