Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD164 Antibody (006), PE, Novus Biologicals™
SDP

Catalog No. NB069656 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB069656 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB069656 Supplier Novus Biologicals Supplier No. NBP290082PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD164 Monoclonal specifically detects CD164 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD164
Applications Flow Cytometry
Classification Monoclonal
Clone 6
Conjugate PE
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD164 antigen, CD164 molecule, sialomucin, Endolyn, MGC-24v, Multi-glycosylated core protein 24, sialomucin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CD164 (Q04900-1; Met1-Asp162).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8763
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.