Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD164 Antibody (67D2), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB002884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002884 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002884 Supplier Novus Biologicals Supplier No. NBP275970C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD164 Monoclonal specifically detects CD164 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface).

Specifications

Antigen CD164
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 67D2
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD164 antigen, CD164 molecule, sialomucin, Endolyn, MGC-24v, Multi-glycosylated core protein 24, sialomucin
Gene Symbols CD164
Host Species Mouse
Immunogen Breast tumor cell line T-47D
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8763
Test Specificity The antibody MEM-154 reacts with an epitope on CD16 antigen that is residing in proximity to FG loop (probably BC or C'E loop). CD16 is a low affinity receptor for aggregated IgG (FcgammaRIII antigen). The antibody MEM-154 reacts with CD16+ granulocytes.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.