Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD173 Antibody (BRIC231), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N1062120AP7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1062120AP7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1062120AP7 Supplier Novus Biologicals Supplier No. NB10062120APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD173 Monoclonal specifically detects CD173 in Human samples. It is validated for Flow Cytometry, Haemagglutination, Immunofluorescence.

Specifications

Antigen CD173
Applications Flow Cytometry, Hemagglutination Assay, Immunofluorescence
Classification Monoclonal
Clone BRIC231
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias H2, Type 2 H blood group antigen
Host Species Mouse
Immunogen HEL cell line
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity NB100-62120 recognises the type 2 H blood group antigen, recently designated CD173. Active H substances in man, are expressed by many cells and tissues and also by erythrocytes
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.