Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD173 Mouse anti-Human, Clone: A46-B/B10, Novus Biologicals™
SDP

Catalog No. NB177817 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177817 20 μg
NB177816 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177817 Supplier Novus Biologicals Supplier No. NBP31393020UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD173 Monoclonal antibody specifically detects CD173 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Hemagglutination Assay, Binding Inhibition

Specifications

Antigen CD173
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Hemagglutination Assay, Inhibition Assays
Classification Monoclonal
Clone A46-B/B10
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry 1-2 ug/ml, Immunofluorescence 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Hemagglutination Assay, Binding Inhibition
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P16442
Gene Alias H2, Type 2 H blood group antigen
Host Species Mouse
Immunogen Human breast cancer MCF-7 cells
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.