Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD177 Antibody (1171A), Biotin, Novus Biologicals™
SDP

Catalog No. NB373482 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373482 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373482 Supplier Novus Biologicals Supplier No. FAB8186B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD177 Monoclonal specifically detects CD177 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD177
Applications Flow Cytometry
Classification Monoclonal
Clone 1171A
Conjugate Biotin
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD177 antigenNB1 GP, CD177 molecule, HNA-2a, HNA2Acell surface receptor, Human neutrophil alloantigen 2a, NB1PRV-1, polycythemia rubra vera 1, Polycythemia rubra vera protein 1, PRV1NB1 glycoprotein
Host Species Rabbit
Immunogen Human embryonic kidney cell line HEK293-derived recombinant mouse CD177, Accession # Q8R2S8
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57126
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.