Learn More
CD18 Mouse anti-Human, FITC, Clone: 6.7, BD
Mouse Monoclonal Antibody
$262.00
Specifications
Antigen | CD18 |
---|---|
Clone | 6.7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB555923
|
BD Biosciences
555923 |
100 Tests |
Each for $262.00
|
|
|||||
BDB557156
|
BD Biosciences
557156 |
50 Tests | N/A | N/A | N/A | ||||
Description
The 6.7 monoclonal antibody specifically binds to β2 integrin, a 90-95 kDa transmembrane protein, which associates non-covalently as a heterodimer with the a chains of CD11a, CD11b, or CD11c. CD18 is expressed on lymphocytes, monocytes, and more weakly on granulocytes. LFA-1 has been shown to play an important role in homotypic and heterotypic cellular adhesion in immune and inflammatory responses.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
CD18 | |
Flow Cytometry | |
FITC | |
RUO | |
Cynomolgus, Human, Pig, Rabbit, Rhesus Monkey | |
Cloned Human DS6 TCR γδ+ Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
6.7 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
ITGB2; Integrin beta-2; Beta subunit for LFA-1, MAC-1/CR3, and p150,95 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.