Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD182 (CXCR2) Monoclonal Antibody (eBio5E8-C7-F10 (5E8-C7-F10)), eBioscience™, Invitrogen™

Catalog No. 14182980
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-182-980 25 μg
50-129-51 100 μg
50-129-52 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-182-980 Supplier Invitrogen™ Supplier No. 14182980
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The eBio5E8-C7-F10 monoclonal antibody reacts with human CD182 (CXCR2, IL-8Rbeta). CD182 is a 67-70 kDa member of the seven-transmembrane spanning G-protein coupled receptor (GPCR) family. CD182 is expressed as a homodimer, or a heterodimer with CD181 (CXCR1, IL-8Ralpha) and is expressed on granulocytes, NK cells, a subset of T cells, mast cells, monocytes, endothelial cells, megakaryocytes and oligodendrocytes. Binding of it's ligands, which include IL-8, NAP-2, GCP-2 and GRO-alpha, -beta and -gamma, induces several biological outcomes such as cell activation, chemotaxis, proliferation and angiogenesis. There are several functional differences between CD181 and CD182. Both receptors are able to mediate chemotaxis and intracellular calcium changes, but only CD181 mediates phospolipase D activation and respiratory burst. Furthermore, studies have shown that IL-8 predomitly mediates its effects on neutrophil function through CD181. Applications Reported: This eBio5E8-C7-F10 (5E8-C7-F10) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio5E8-C7-F10 (5E8-C7-F10) antibody has been tested by flow cytometric analysis of normal human peripheral blood. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.

CXCR2 is a receptor for interleukin-8, which is a powerful neutrophil chemotactic factor. It is a member of the GPCR family. Binding of IL-8 to the receptor causes activation of neutrophils. This response is mediated via a G-protein that activate a phosphatidylinositol-calcium second messenger system. This receptor binds to IL-8 with a high affinity and to GRO/MGSA and NAP-2 also with a high affinity. It has been reported to be expressed in a wide variety of tissues. ESTs have been isolated from human placenta and thymus libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD182 (CXCR2)
Applications Flow Cytometry
Classification Monoclonal
Clone eBio5E8-C7-F10 (5E8-C7-F10)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CXCR2
Gene Accession No. P25025
Gene Alias C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD128; CD182; CD182 antigen; CDw128; CDw128b; chemokine (C-X-C motif) receptor 2; chemokine (CXC) receptor 2; chemokine (C-X-C) receptor 2; Cmkar2; CXC; C-X-C chemokine receptor type 2; CXC motif chemokine; C-X-C motif chemokine receptor 2; Cxcr2; CXC-R2; CXCR-2; CXCR2 gene for IL8 receptor type B; Gpcr16; GRO/MGSA receptor; high affinity interleukin-8 receptor B; IL-8 receptor alpha chain; Il-8 receptor b; IL-8 receptor beta; IL-8 receptor type 2; IL-8R B; IL8R2; IL8RA; Il8rb; IL-8rb; IL-8Rh; interleukin 8 receptor B; interleukin 8 receptor type 2; interleukin 8 receptor, beta; interleukin-8 receptor type B; mIL-8RH
Gene Symbols CXCR2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3579
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.